Alfa Aesar™ Methyl 2-chloro-3-hydroxypropionate, 97%

Manufacturer: Alfa Aesar™ H2762306

Quantity 5g

 View more versions of this product

Catalog No. AAH2762306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98070-39-4
    Molecular Formula C4H7ClO3
    Formula Weight 138.55
    MDL Number MFCD07368971
    Synonym 2-Chloro-3-hydroxypropionic acid methyl ester
    Chemical Name or Material Methyl 2-chloro-3-hydroxypropionate

    Specifications

    Formula Weight 138.55
    Molecular Formula C4H7ClO3
    MDL Number MFCD07368971
    CAS 98070-39-4
    Synonym 2-Chloro-3-hydroxypropionic acid methyl ester
    Health Hazard 1 Danger
    Boiling Point 105° to 109°C (20mmHg)
    Quantity 5g
    Flash Point 110°C (230°F)
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Methyl 2-chloro-3-hydroxypropionate
    Assay 97%

    Description

    Applications
    Treatment of 3-aryl-2-hydroxypropanoic esters which is a derivative of methyl 2-chloro-3-hydroxypropionate with diethylaminosulfur trifluoride (DAST) gives considerable amounts of rearranged 2-aryl-3-fluoropropanoic esters together with the expected products.

    Notes
    Store at 2-8°C. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit