Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
175137-27-6
C7H4ClF3N2O2
240.57
VLUBJYUOPNGBOQ-UHFFFAOYSA-N
methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate,methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate,methylchlorotrifluoromethylpyrimidinecarboxylate,methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate,acmc-20aifi,pubchem10174,2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine,methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca,2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine,2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
3696469
methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
COC(=O)C1=CN=C(N=C1C(F)(F)F)Cl
175137-27-6
C7H4ClF3N2O2
240.57
VLUBJYUOPNGBOQ-UHFFFAOYSA-N
methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate,methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate,methylchlorotrifluoromethylpyrimidinecarboxylate,methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate,acmc-20aifi,pubchem10174,2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine,methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca,2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine,2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
3696469
methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
COC(=O)C1=CN=C(N=C1C(F)(F)F)Cl
Boiling Point 56.0°C (0.2 mmHg)
Quantity 5g
Infrared Spectrum Authentic
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate

GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

Warning