Alfa Aesar™ Methyl 2-chloro-6-methylpyridine-4-carboxylate, 97%

Manufacturer: Alfa Aesar™ H56006MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH56006MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3998-90-1
    Molecular Formula C8H8ClNO2
    Formula Weight 185.61
    MDL Number MFCD04038391
    Synonym 2-Chloro-6-methylpyridine-4-carboxylic acid methyl ester; Methyl 2-chloro-6-methylisonicotinate
    Chemical Name or Material Methyl 2-chloro-6-methylpyridine-4-carboxylate

    Specifications

    Formula Weight 185.61
    MDL Number MFCD04038391
    Melting Point 58° to 62°C
    Quantity 250mg
    Molecular Formula C8H8ClNO2
    CAS 3998-90-1
    Synonym 2-Chloro-6-methylpyridine-4-carboxylic acid methyl ester; Methyl 2-chloro-6-methylisonicotinate
    Health Hazard 1 Danger
    Boiling Point 124° to 125°C (7mmHg)
    Health Hazard 2 GHS H Statement
    H318-H317-H335-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Methyl 2-chloro-6-methylpyridine-4-carboxylate
    Assay 97%

    GHS H Statement
    H318-H317-H335-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit