Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-chlorobenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
610-96-8
C8H7ClO2
170.592
MFCD00016337
JAVRNIFMYIJXIE-UHFFFAOYSA-N
methyl o-chlorobenzoate,methyl-2-chlorobenzoate,benzoic acid, 2-chloro-, methyl ester,2-chlorobenzoic acid methyl ester,unii-g344nes07r,benzoic acid, o-chloro-, methyl ester,benzoicacid, 2-chloro-, methyl ester,pubchem3707,methyl-o-chlorobenzoate,acmc-209moz
11895
methyl 2-chlorobenzoate
COC(=O)C1=CC=CC=C1Cl
610-96-8
C8H7ClO2
170.592
MFCD00016337
JAVRNIFMYIJXIE-UHFFFAOYSA-N
methyl o-chlorobenzoate,methyl-2-chlorobenzoate,benzoic acid, 2-chloro-, methyl ester,2-chlorobenzoic acid methyl ester,unii-g344nes07r,benzoic acid, o-chloro-, methyl ester,benzoicacid, 2-chloro-, methyl ester,pubchem3707,methyl-o-chlorobenzoate,acmc-209moz
11895
methyl 2-chlorobenzoate
COC(=O)C1=CC=CC=C1Cl
Specific Gravity 1.191
Refractive Index 1.5351 to 1.5370
Quantity 25g
Packaging Glass bottle
Color Colorless
Density 1.1910g/mL
Flash Point 108°C
Infrared Spectrum Authentic
Boiling Point 233.0°C to 235.0°C
Assay Percent Range 98%
Formula Weight 170.59
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl 2-chlorobenzoate, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-242-0

TSCA : TSCA