Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-chloropyridine-3-carboxylate, 98%

Click to view available options
Quantity:
5g
25g
100g
40134-18-7
C7H6ClNO2
171.58
MFCD01318629
MYGAJZBZLONIBZ-UHFFFAOYSA-N
methyl 2-chloronicotinate,2-chloronicotinic acid methyl ester,2-chloro-3-pyridinecarboxylic acid methyl ester,methyl2-chloronicotinate,methyl 2-chloro-3-pyridinecarboxylate,3-pyridinecarboxylic acid, 2-chloro-, methyl ester,2-chloro-nicotinic acid methyl ester,methyl chloronicotinate,pubchem15917,acmc-1am6z
819931
methyl 2-chloropyridine-3-carboxylate
COC(=O)C1=C(N=CC=C1)Cl
40134-18-7
C7H6ClNO2
171.58
MFCD01318629
MYGAJZBZLONIBZ-UHFFFAOYSA-N
methyl 2-chloronicotinate,2-chloronicotinic acid methyl ester,2-chloro-3-pyridinecarboxylic acid methyl ester,methyl2-chloronicotinate,methyl 2-chloro-3-pyridinecarboxylate,3-pyridinecarboxylic acid, 2-chloro-, methyl ester,2-chloro-nicotinic acid methyl ester,methyl chloronicotinate,pubchem15917,acmc-1am6z
819931
methyl 2-chloropyridine-3-carboxylate
COC(=O)C1=C(N=CC=C1)Cl
Color Colorless
Density 1.314g/mL
Refractive Index 1.5370
Boiling Point 150°C (1mmHg)
Quantity 25g
Beilstein 122129
Formula Weight 171.58
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl 2-chloropyridine-3-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning