Alfa Aesar™ Methyl 2-(chlorosulfonyl)benzoate, 94%

Manufacturer: Alfa Aesar™ B2321114

Quantity 25g

 View more versions of this product

Catalog No. AAB2321114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26638-43-7
    Molecular Formula C8H7ClO4S
    Formula Weight 234.66
    MDL Number MFCD00009797
    UN Number UN3261
    Synonym 2-(Chlorosulfonyl)benzoic acid methyl ester; 2-(Methoxycarbonyl)benzenesulfonyl chloride
    Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate

    Specifications

    Formula Weight 234.66
    TSCA TSCA
    Molecular Formula C8H7ClO4S
    MDL Number MFCD00009797
    CAS 26638-43-7
    UN Number UN3261
    Synonym 2-(Chlorosulfonyl)benzoic acid methyl ester; 2-(Methoxycarbonyl)benzenesulfonyl chloride
    Melting Point 59° to 64°C
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate
    Assay 94%

    Description

    Applications
    Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

    Solubility
    Reacts with water.

    Notes
    Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2° to 8°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit