Alfa Aesar™ Methyl 2-diphenylphosphino-1-naphthoate, 98%

Manufacturer: Alfa Aesar™ H3092814

Quantity 25g

 View more versions of this product

Catalog No. AAH3092814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 178176-78-8
    Molecular Formula C24H19O2P
    Formula Weight 370.39
    MDL Number MFCD12546191
    Synonym 2-Diphenylphosphino-1-naphthoic acid methyl ester
    Chemical Name or Material Methyl 2-diphenylphosphino-1-naphthoate

    Specifications

    Formula Weight 370.39
    MDL Number MFCD12546191
    Health Hazard 1 Warning
    Melting Point 107° to 108°C
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C24H19O2P
    CAS 178176-78-8
    Synonym 2-Diphenylphosphino-1-naphthoic acid methyl ester
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Methyl 2-diphenylphosphino-1-naphthoate
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit