Alfa Aesar™ Methyl 2-fluoro-6-methoxybenzoate, 98%

Manufacturer: Alfa Aesar™ H3295509

Quantity 10g

 View more versions of this product

Catalog No. AAH3295509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 178747-79-0
    Molecular Formula C9H9FO3
    Formula Weight 184.17
    MDL Number MFCD03788537
    Synonym 2-Fluoro-6-methoxybenzoic acid methyl ester
    Chemical Name or Material Methyl 2-fluoro-6-methoxybenzoate

    Specifications

    Formula Weight 184.17
    Molecular Formula C9H9FO3
    MDL Number MFCD03788537
    CAS 178747-79-0
    Synonym 2-Fluoro-6-methoxybenzoic acid methyl ester
    Quantity 10g
    Chemical Name or Material Methyl 2-fluoro-6-methoxybenzoate
    Assay 98%

    Description

    Applications
    Methyl 2-Fluoro-6-methoxybenzoate is used in the synthetic preparation of thiazinotrienomycin E which is an ansamycin macrolactam antibiotic. Useful intermediates for the synthesis of thiazolones and 2,4,5-trisubstituted thiazoles.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit