Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl (2-furfurylthio)acetate, 97%

Click to view available options
Quantity:
5g
25g
100g
108499-33-8
C8H10O3S
186.225
MFCD02283460
XPVNCXPHDABNLE-UHFFFAOYSA-N
methyl 2-furfurylthio acetate,methyl 2-furan-2-ylmethyl thio acetate,acetic acid,2-2-furanylmethyl thio-, methyl ester,methyl 2-furan-2-ylmethyl sulfanyl acetate,acmc-20f6v3,aceticacid, 2-furanylmethyl thio-methylester,methyl-2-furfurylthio acetate,methyl 2-2-furylmethylthio acetate,furfurylthio acetic acid methyl ester,2-furfurylthio acetic acid methyl ester
7022533
methyl 2-(furan-2-ylmethylsulfanyl)acetate
COC(=O)CSCC1=CC=CO1
108499-33-8
C8H10O3S
186.225
MFCD02283460
XPVNCXPHDABNLE-UHFFFAOYSA-N
methyl 2-furfurylthio acetate,methyl 2-furan-2-ylmethyl thio acetate,acetic acid,2-2-furanylmethyl thio-, methyl ester,methyl 2-furan-2-ylmethyl sulfanyl acetate,acmc-20f6v3,aceticacid, 2-furanylmethyl thio-methylester,methyl-2-furfurylthio acetate,methyl 2-2-furylmethylthio acetate,furfurylthio acetic acid methyl ester,2-furfurylthio acetic acid methyl ester
7022533
methyl 2-(furan-2-ylmethylsulfanyl)acetate
COC(=O)CSCC1=CC=CO1
Boiling Point 95°C (3mmHg)
Quantity 25g
Flash Point >100°C (212°F)
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Methyl (2-furfurylthio)acetate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning