Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-furoate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
611-13-2
C6H6O3
126.11
MFCD00003236
HDJLSECJEQSPKW-UHFFFAOYSA-N
methyl 2-furoate,methyl pyromucate,methyl 2-furancarboxylate,methyl furoate,2-furancarboxylic acid, methyl ester,2-methoxycarbonyl furan,pyromucic acid methyl ester,methyl 2-furylcarboxylate,2-furoic acid, methyl ester,furoic acid, methyl ester
11902
methyl furan-2-carboxylate
COC(=O)C1=CC=CO1
611-13-2
C6H6O3
126.11
MFCD00003236
HDJLSECJEQSPKW-UHFFFAOYSA-N
methyl 2-furoate,methyl pyromucate,methyl 2-furancarboxylate,methyl furoate,2-furancarboxylic acid, methyl ester,2-methoxycarbonyl furan,pyromucic acid methyl ester,methyl 2-furylcarboxylate,2-furoic acid, methyl ester,furoic acid, methyl ester
11902
methyl furan-2-carboxylate
COC(=O)C1=CC=CO1
Color Yellow to Brown
Infrared Spectrum Authentic
Boiling Point 181.0°C
Quantity 25g
Density 1.1760g/mL
Refractive Index 1.4830 to 1.4890
Packaging Glass bottle
Specific Gravity 1.176
Flash Point 73°C
Assay Percent Range 97.5% min. (GC)
Merck Index 15,6149
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 126.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl 2-furoate, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-254-6

RTECSNumber : LV1950000

TSCA : TSCA