Alfa Aesar™ Methyl 2-hexynoate, 98+%

Manufacturer: Alfa Aesar™ B2306514

Quantity 25g

 View more versions of this product

Catalog No. AAB2306514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18937-79-6
    Molecular Formula CH3CH2CH2C≡CCO2CH3
    Formula Weight 126.16
    MDL Number MFCD00041642
    Beilstein 1750018
    Synonym 2-Hexynoic acid methyl ester
    Chemical Name or Material Methyl 2-hexynoate

    Specifications

    Formula Weight 126.16
    Molecular Formula CH3CH2CH2C≡CCO2CH3
    MDL Number MFCD00041642
    CAS 18937-79-6
    Beilstein 1750018
    Synonym 2-Hexynoic acid methyl ester
    Health Hazard 1 Warning
    Recommended Storage 2° to 8°C
    Flash Point 67°C (152°F)
    Boiling Point 64° to 66°C (10mmHg)
    Quantity 25g
    Chemical Name or Material Methyl 2-hexynoate
    Assay ≥98%

    2° to 8°C

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit