Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-hexynoate, 98+%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
18937-79-6
C7H10O2
126.15
MFCD00041642
DHZPZBFOJDXEHS-UHFFFAOYSA-N
methyl 2-hexynoate,2-hexynoic acid methyl ester,2-hexynoic acid, methyl ester,hex-2-ynoic acid methyl ester,methyl hexynoate,acmc-20akik,2-hexynoicacid,methyl ester,2-hexynoicacid, methyl ester
87860
methyl hex-2-ynoate
CCCC#CC(=O)OC
18937-79-6
C7H10O2
126.15
MFCD00041642
DHZPZBFOJDXEHS-UHFFFAOYSA-N
methyl 2-hexynoate,2-hexynoic acid methyl ester,2-hexynoic acid, methyl ester,hex-2-ynoic acid methyl ester,methyl hexynoate,acmc-20akik,2-hexynoicacid,methyl ester,2-hexynoicacid, methyl ester
87860
methyl hex-2-ynoate
CCCC#CC(=O)OC
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Density 0.9430g/mL
Specific Gravity 0.943
Flash Point 67°C
Boiling Point 64.0°C to 66.0°C (10.0mmHg)
Quantity 25g
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Linear Formula CH3(CH2)2C≡CCOOCH3
Formula Weight 126.15
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material Methyl 2-hexynoate
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.