Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-hydroxy-3-nitrobenzoate, 98%

Click to view available options
Quantity:
1g
5g
25g
22621-41-6
C8H7NO5
197.146
MFCD00272263
NIBVYEHAFBEVFI-UHFFFAOYSA-N
methyl 2-hydroxy-3-nitrobenzoate,methyl 3-nitrosalicylate,benzoic acid, 2-hydroxy-3-nitro-, methyl ester,2-hydroxy-3-nitrobenzoic acid methyl ester,2-hydroxy-3-nitro-benzoic acid methyl ester,salicylic acid, 3-nitro-, methyl ester,methyl-3-nitrosalicylat,methyl-m-nitrosalicylate,acmc-209fxo,ksc490c4l
596904
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
22621-41-6
C8H7NO5
197.146
MFCD00272263
NIBVYEHAFBEVFI-UHFFFAOYSA-N
methyl 2-hydroxy-3-nitrobenzoate,methyl 3-nitrosalicylate,benzoic acid, 2-hydroxy-3-nitro-, methyl ester,2-hydroxy-3-nitrobenzoic acid methyl ester,2-hydroxy-3-nitro-benzoic acid methyl ester,salicylic acid, 3-nitro-, methyl ester,methyl-3-nitrosalicylat,methyl-m-nitrosalicylate,acmc-209fxo,ksc490c4l
596904
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
Quantity 5g
Melting Point 129°C to 132°C
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-3-nitrobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning