Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-hydroxy-5-nitrobenzoate, 98%

Click to view available options
Quantity:
10g
50g
250g
17302-46-4
C8H7NO5
197.146
MFCD00024695
UUBFELFUKFJSRD-UHFFFAOYSA-N
methyl 2-hydroxy-5-nitrobenzoate,methyl 5-nitrosalicylate,methyl-5-nitrosalicylate,5-nitrosalicylic acid methyl ester,benzoic acid, 2-hydroxy-5-nitro-, methyl ester,2-hydroxy-5-nitrobenzoic acid methyl ester,methyl 5-nitro-salicylate,acmc-20a62q,methyl 5-nitro-2-hydroxybenzoate,methyl-2-hydroxy-5-nitrobenzoate
87041
COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O
17302-46-4
C8H7NO5
197.146
MFCD00024695
UUBFELFUKFJSRD-UHFFFAOYSA-N
methyl 2-hydroxy-5-nitrobenzoate,methyl 5-nitrosalicylate,methyl-5-nitrosalicylate,5-nitrosalicylic acid methyl ester,benzoic acid, 2-hydroxy-5-nitro-, methyl ester,2-hydroxy-5-nitrobenzoic acid methyl ester,methyl 5-nitro-salicylate,acmc-20a62q,methyl 5-nitro-2-hydroxybenzoate,methyl-2-hydroxy-5-nitrobenzoate
87041
COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O
Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 10g
Melting Point 114°C to 117°C
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-5-nitrobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning