Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-hydroxyisobutyrate 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
2110-78-3
C5H10O3
118.13
MFCD00004457
XYVQFUJDGOBPQI-UHFFFAOYSA-N
methyl 2-hydroxyisobutyrate,methyl 2-methyllactate,2-hydroxyisobutyric acid methyl ester,methyl 2-hydroxy-2-methylpropionate,methyl2-hydroxyisobutyrate,propanoic acid, 2-hydroxy-2-methyl-, methyl ester,methyl alpha-hydroxyisobutyrate,methyl .alpha.-hydroxyisobutyrate,methyl ester of 2-methyllactic acid,methyl 2-hydroxy-2-methyl-propanoate
75024
methyl 2-hydroxy-2-methylpropanoate
CC(C)(C(=O)OC)O
2110-78-3
C5H10O3
118.13
MFCD00004457
XYVQFUJDGOBPQI-UHFFFAOYSA-N
methyl 2-hydroxyisobutyrate,methyl 2-methyllactate,2-hydroxyisobutyric acid methyl ester,methyl 2-hydroxy-2-methylpropionate,methyl2-hydroxyisobutyrate,propanoic acid, 2-hydroxy-2-methyl-, methyl ester,methyl alpha-hydroxyisobutyrate,methyl .alpha.-hydroxyisobutyrate,methyl ester of 2-methyllactic acid,methyl 2-hydroxy-2-methyl-propanoate
75024
methyl 2-hydroxy-2-methylpropanoate
CC(C)(C(=O)OC)O
Density 1.0300g/mL
Refractive Index 1.4100 to 1.412
Boiling Point 137°C
Flash Point 41°C
Specific Gravity 1.03
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Quantity 50g
Assay Percent Range 99%
Linear Formula (CH3)2C(OH)CO2CH3
Beilstein 03, II, 223
Formula Weight 118.13
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Methyl 2-hydroxyisobutyrate, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-301-2

TSCA : TSCA