Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-methoxy-5-nitrobenzoate, 98%

Click to view available options
Quantity:
5g
25g
34841-11-7
C9H9NO5
211.173
MFCD00673023
DOBFJVVTBNTGCW-UHFFFAOYSA-N
2-methoxy-5-nitrobenzoic acid methyl ester,methyl2-methoxy-5-nitrobenzoate,methyl-2-methoxy-5-nitrobenzoate,5-nitro-o-anisic acid methyl ester,pubchem14052,acmc-1adrt,methyl 5-nitro-o-anisate,asischem d13383,rarechem al bf 0877,timtec-bb sbb014757
7020925
methyl 2-methoxy-5-nitrobenzoate
COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC
34841-11-7
C9H9NO5
211.173
MFCD00673023
DOBFJVVTBNTGCW-UHFFFAOYSA-N
2-methoxy-5-nitrobenzoic acid methyl ester,methyl2-methoxy-5-nitrobenzoate,methyl-2-methoxy-5-nitrobenzoate,5-nitro-o-anisic acid methyl ester,pubchem14052,acmc-1adrt,methyl 5-nitro-o-anisate,asischem d13383,rarechem al bf 0877,timtec-bb sbb014757
7020925
methyl 2-methoxy-5-nitrobenzoate
COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC
Melting Point 99°C to 102°C
Quantity 25g
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material Methyl 2-methoxy-5-nitrobenzoate