Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-methoxybenzoate, 98+%

Click to view available options
Quantity:
25g
100g
606-45-1
C9H10O3
166.176
MFCD00008423
PFYHAAAQPNMZHO-UHFFFAOYSA-N
methyl o-anisate,dimethyl salicylate,o-anisic acid, methyl ester,methyl o-methoxybenzoate,o-methoxy methyl benzoate,o-methoxybenzoic acid methyl ester,methylsalicylate methyl ether,2-methoxybenzoic acid methyl ester,benzoic acid, 2-methoxy-, methyl ester,2-methoxybenzoic acid, methyl ester
61151
methyl 2-methoxybenzoate
COC1=CC=CC=C1C(=O)OC
606-45-1
C9H10O3
166.176
MFCD00008423
PFYHAAAQPNMZHO-UHFFFAOYSA-N
methyl o-anisate,dimethyl salicylate,o-anisic acid, methyl ester,methyl o-methoxybenzoate,o-methoxy methyl benzoate,o-methoxybenzoic acid methyl ester,methylsalicylate methyl ether,2-methoxybenzoic acid methyl ester,benzoic acid, 2-methoxy-, methyl ester,2-methoxybenzoic acid, methyl ester
61151
methyl 2-methoxybenzoate
COC1=CC=CC=C1C(=O)OC
Refractive Index 1.534
Density 1.16
Boiling Point 246°C to 248°C
Quantity 100g
Odor Characteristic
Flash Point >110°C (230°F)
Beilstein 2207356
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Methyl 2-methoxybenzoate

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-118-6

RTECSNumber : DH3549000

TSCA : Yes

Recommended Storage : Ambient temperatures