Alfa Aesar™ Methyl 2-methyl-3-furoate, 97%

Manufacturer: Alfa Aesar™ A1197922

Quantity 100g

 View more versions of this product

Catalog No. AAA1197922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6141-58-8
    Molecular Formula C7H8O3
    Formula Weight 140.14
    MDL Number MFCD00003249
    Beilstein 1342040
    Synonym 2-Methylfuran-3-carboxylic acid methyl ester; 2-Methyl-3-furoic acid methyl ester
    Chemical Name or Material Methyl 2-methyl-3-furoate

    Specifications

    Formula Weight 140.14
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 6141-58-8
    MDL Number MFCD00003249
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Beilstein 1342040
    Synonym 2-Methylfuran-3-carboxylic acid methyl ester; 2-Methyl-3-furoic acid methyl ester
    Molecular Formula C7H8O3
    Boiling Point 71° to 72°C (20mmHg)
    Flash Point 63°C (145°F)
    Quantity 100g
    Chemical Name or Material Methyl 2-methyl-3-furoate
    Assay 97%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit