Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-methylbutyrate, 98%, Acros Organics

Click to view available options
Quantity:
100g
1kg
5kg
868-57-5
C6H12O2
116.16
OCWLYWIFNDCWRZ-UHFFFAOYSA-N
methyl 2-methylbutyrate,butanoic acid, 2-methyl-, methyl ester,methyl 2-methyl butyrate,methyl s-2-methylbutanoate,2-methylbutyric acid methyl ester,butyric acid, 2-methyl-, methyl ester,dl-2-methylbutyric acid methyl ester,methyl dl-2-methylbutyrate,methyl alpha-methylbutyrate,methyl alpha-methylbutanoate
13357
methyl 2-methylbutanoate
CCC(C)C(=O)OC
868-57-5
C6H12O2
116.16
OCWLYWIFNDCWRZ-UHFFFAOYSA-N
methyl 2-methylbutyrate,butanoic acid, 2-methyl-, methyl ester,methyl 2-methyl butyrate,methyl s-2-methylbutanoate,2-methylbutyric acid methyl ester,butyric acid, 2-methyl-, methyl ester,dl-2-methylbutyric acid methyl ester,methyl dl-2-methylbutyrate,methyl alpha-methylbutyrate,methyl alpha-methylbutanoate
13357
methyl 2-methylbutanoate
CCC(C)C(=O)OC
Flash Point 18°C
Packaging Glass bottle
Quantity 100g
Density 0.8850g/mL
Boiling Point 115.0°C
Specific Gravity 0.885
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH2CH(CH3)COOCH3
Solubility Information Solubility in water: practically insoluble
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Methyl 2-methylbutyrate
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS Signal Word: Danger

EINECSNumber : 212-778-