Alfa Aesar™ Methyl 2-(methylthio)butyrate, 98%

Manufacturer: Alfa Aesar™ B2230314

Quantity 25g

 View more versions of this product

Catalog No. AAB2230314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51534-66-8
    Molecular Formula C6H12O2S
    Formula Weight 148.23
    MDL Number MFCD00013393
    Beilstein 1925141
    Synonym 2-(Methylthio)butyric acid methyl ester; Methyl 2-(methylmercapto)butyrate
    Chemical Name or Material Methyl 2-(methylthio)butyrate

    Specifications

    Formula Weight 148.23
    Molecular Formula C6H12O2S
    MDL Number MFCD00013393
    Health Hazard 1 Warning
    Boiling Point 103° to 104°C (35mmHg)
    Quantity 25g
    Flash Point 64°C (147°F)
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 51534-66-8
    Beilstein 1925141
    Synonym 2-(Methylthio)butyric acid methyl ester; Methyl 2-(methylmercapto)butyrate
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Chemical Name or Material Methyl 2-(methylthio)butyrate
    Assay 98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit