Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl 2-(methylthio)butyrate, 98%
Click to view available options
Quantity:
5g
25g
Specifications
Boiling Point | 103°C to 104°C (35mmHg) |
Flash Point | 64°C (147°F) |
Refractive Index | 1.461 |
Density | 1.028 |
Quantity | 5g |
Beilstein | 1925141 |
Formula Weight | 148.23 |
Percent Purity | 98% |
Chemical Name or Material | Methyl 2-(methylthio)butyrate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
TSCA : No
Recommended Storage : Ambient temperatures