Alfa Aesar™ Methyl 2-(methylthio)propionate, 99%

Manufacturer: Alfa Aesar™ B2197914

Quantity 25g

 View more versions of this product

Catalog No. AAB2197914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61366-76-5
    Molecular Formula C5H10O2S
    Formula Weight 134.2
    MDL Number MFCD02683079
    Synonym Methyl 2-(methylmercapto)propionate; 2-(Methylthio)propionic acid methyl ester
    Chemical Name or Material Methyl 2-(methylthio)propionate

    Specifications

    Formula Weight 134.2
    Molecular Formula C5H10O2S
    MDL Number MFCD02683079
    CAS 61366-76-5
    Synonym Methyl 2-(methylmercapto)propionate; 2-(Methylthio)propionic acid methyl ester
    Quantity 25g
    Chemical Name or Material Methyl 2-(methylthio)propionate
    Assay 99%

    Description

    Applications
    Methyl 2-(methylthio)propionate is used in the contribution of overall aroma perception of the ripe fruit.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit