Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl 2-(methylthio)propionate, 99%
Click to view available options
Quantity:
5g
25g
Description
Applications
Methyl 2-(methylthio)propionate is used in the contribution of overall aroma perception of the ripe fruit.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Methyl 2-(methylthio)propionate is used in the contribution of overall aroma perception of the ripe fruit.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Refractive Index | 1.462 |
Quantity | 5g |
Solubility Information | Immiscible with water. |
Formula Weight | 134.2 |
Percent Purity | 99% |
Chemical Name or Material | Methyl 2-(methylthio)propionate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures