Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-methylthiopyrimidine-5-carboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
5g
38275-41-1
C7H8N2O2S
184.22
PSYRMEZGAWNWHV-UHFFFAOYSA-N
methyl 2-methylthio pyrimidine-5-carboxylate,methyl 2-methylsulfanyl pyrimidine-5-carboxylate,methyl 2-methylthiopyrimidine-5-carboxylate,methyl 2-methylthio-5-pyrimidinecarboxylate,2-methylthio-pyrimidine-5-carboxylic acid methyl ester,5-pyrimidinecarboxylic acid, 2-methylthio-, methyl ester,methyl2-methylthio pyrimidine-5-carboxylate,methyl 2-methylthio-5-pyrimidinearboxylate,methyl-2-methylthio pyrimidine-5-carboxylate,methyl 2-methylsulphanyl pyrimidine-5-carboxylate
11194721
methyl 2-methylsulfanylpyrimidine-5-carboxylate
COC(=O)C1=CN=C(N=C1)SC
38275-41-1
C7H8N2O2S
184.22
PSYRMEZGAWNWHV-UHFFFAOYSA-N
methyl 2-methylthio pyrimidine-5-carboxylate,methyl 2-methylsulfanyl pyrimidine-5-carboxylate,methyl 2-methylthiopyrimidine-5-carboxylate,methyl 2-methylthio-5-pyrimidinecarboxylate,2-methylthio-pyrimidine-5-carboxylic acid methyl ester,5-pyrimidinecarboxylic acid, 2-methylthio-, methyl ester,methyl2-methylthio pyrimidine-5-carboxylate,methyl 2-methylthio-5-pyrimidinearboxylate,methyl-2-methylthio pyrimidine-5-carboxylate,methyl 2-methylsulphanyl pyrimidine-5-carboxylate
11194721
methyl 2-methylsulfanylpyrimidine-5-carboxylate
COC(=O)C1=CN=C(N=C1)SC
Quantity 5g
Infrared Spectrum Authentic
Melting Point 96.0°C to 97.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 184.22
Percent Purity 97%
Chemical Name or Material Methyl 2-methylthiopyrimidine-5-carboxylate

GHS H Statement
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning