Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-nitrobenzoate, 98+%

Click to view available options
Quantity:
25g
100g
500g
606-27-9
C8H7NO4
181.147
MFCD00007136
AOXPHVNMBPFOFS-UHFFFAOYSA-N
methyl o-nitrobenzoate,benzoic acid, 2-nitro-, methyl ester,2-nitro-benzoic acid methyl ester,benzoic acid, o-nitro-, methyl ester,2-nitrobenzoic acid methyl ester,methyl-2-nitrobenzoate,methyl o-nitro benzoate,methyl ortho-nitrobenzoate,acmc-209ml4,nitrobenzoic acid methyl ester
69072
methyl 2-nitrobenzoate
COC(=O)C1=CC=CC=C1[N+](=O)[O-]
606-27-9
C8H7NO4
181.147
MFCD00007136
AOXPHVNMBPFOFS-UHFFFAOYSA-N
methyl o-nitrobenzoate,benzoic acid, 2-nitro-, methyl ester,2-nitro-benzoic acid methyl ester,benzoic acid, o-nitro-, methyl ester,2-nitrobenzoic acid methyl ester,methyl-2-nitrobenzoate,methyl o-nitro benzoate,methyl ortho-nitrobenzoate,acmc-209ml4,nitrobenzoic acid methyl ester
69072
methyl 2-nitrobenzoate
COC(=O)C1=CC=CC=C1[N+](=O)[O-]
Melting Point -13°C
Quantity 500g
Flash Point >110°C (230°F)
Boiling Point 275°C
Beilstein 2050101
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material Methyl 2-nitrobenzoate