Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-oxocyclohexanecarboxylate, 90%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
41302-34-5
C8H12O3
156.2
MFCD00077699
JEENWEAPRWGXSG-UHFFFAOYSA-N
methyl 2-oxocyclohexanecarboxylate,2-methoxycarbonylcyclohexanone,2-carbomethoxycyclohexanone,cyclohexanecarboxylic acid, 2-oxo-, methyl ester,methyl cyclohexanone-2-carboxylate,2-oxo-cyclohexanecarboxylic acid methyl ester,2-hydroxy methyl cyclohexanone,2-oxocyclohexanecarboxylic acid, methyl ester,pubchem20380,acmc-1ao3u
170489
methyl 2-oxocyclohexane-1-carboxylate
COC(=O)C1CCCCC1=O
41302-34-5
C8H12O3
156.2
MFCD00077699
JEENWEAPRWGXSG-UHFFFAOYSA-N
methyl 2-oxocyclohexanecarboxylate,2-methoxycarbonylcyclohexanone,2-carbomethoxycyclohexanone,cyclohexanecarboxylic acid, 2-oxo-, methyl ester,methyl cyclohexanone-2-carboxylate,2-oxo-cyclohexanecarboxylic acid methyl ester,2-hydroxy methyl cyclohexanone,2-oxocyclohexanecarboxylic acid, methyl ester,pubchem20380,acmc-1ao3u
170489
methyl 2-oxocyclohexane-1-carboxylate
COC(=O)C1CCCCC1=O
Boiling Point 50°C (0.2mmHg)
Refractive Index 1.4830 to 1.486
Flash Point 85°C
Specific Gravity 1.109
Density 1.1090g/mL
Packaging Glass bottle
Color Colorless to Yellow
Quantity 50mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Beilstein 10, IV, 2606
Formula Weight 156.2
Physical Form Liquid
Percent Purity ≥88% may occur as tautomers
Chemical Name or Material Methyl 2-oxocyclohexanecarboxylate
Show More Show Less