Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl 2-oxocyclopentanecarboxylate, 97%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Density | 1.145 |
Refractive Index | 1.4565 |
Quantity | 5g |
Flash Point | >110°C (230°F) |
Boiling Point | 105°C to 107°C (20mmHg) |
Formula Weight | 142.16 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-oxocyclopentanecarboxylate |
Safety and Handling
P210-P233-P235-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
H227-H302-H315-H319
EINECSNumber : 233-962-7
TSCA : Yes
Recommended Storage : Ambient temperatures