Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-oxoindoline-7-carboxylate, 96%

Click to view available options
Quantity:
250mg
1g
380427-39-4
C10H9NO3
191.186
MFCD02179610
XVJRNLIMSQFUAI-UHFFFAOYSA-N
methyl 2-oxoindoline-7-carboxylate,methyl oxindole-7-carboxylate,methyl 2-oxo-2,3-dihydro-1h-indole-7-carboxylate,1h-indole-7-carboxylic acid, 2,3-dihydro-2-oxo-, methyl ester,acmc-1aje4,methyl oxindole-7-carboxylate,,methyl 2-oxindole-7-carboxylate,2-oxyindole-7-carboxylic acid methyl ester,1h-indole-7-carboxylicacid, 2,3-dihydro-2-oxo-, methyl ester
2773518
methyl 2-oxo-1,3-dihydroindole-7-carboxylate
COC(=O)C1=C2C(=CC=C1)CC(=O)N2
380427-39-4
C10H9NO3
191.186
MFCD02179610
XVJRNLIMSQFUAI-UHFFFAOYSA-N
methyl 2-oxoindoline-7-carboxylate,methyl oxindole-7-carboxylate,methyl 2-oxo-2,3-dihydro-1h-indole-7-carboxylate,1h-indole-7-carboxylic acid, 2,3-dihydro-2-oxo-, methyl ester,acmc-1aje4,methyl oxindole-7-carboxylate,,methyl 2-oxindole-7-carboxylate,2-oxyindole-7-carboxylic acid methyl ester,1h-indole-7-carboxylicacid, 2,3-dihydro-2-oxo-, methyl ester
2773518
methyl 2-oxo-1,3-dihydroindole-7-carboxylate
COC(=O)C1=C2C(=CC=C1)CC(=O)N2
Quantity 250mg
Beilstein 15864880
Formula Weight 191.19
Physical Form Solid
Percent Purity 96%
Chemical Name or Material Methyl 2-oxoindoline-7-carboxylate