Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-pyridylacetate 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1658-42-0
C8H9NO2
151.165
MFCD00006358
ORAKNQSHWMHCEY-UHFFFAOYSA-N
methyl 2-pyridylacetate,methyl 2-pyridin-2-yl acetate,2-pyridylacetic acid methyl ester,2-pyridineacetic acid, methyl ester,methyl 2-pyridinylacetate,methyl pyridin-2-ylacetate,methyl 2-pyridineacetate,2-pyridineacetic acid methyl ester,pyridin-2-yl-acetic acid methyl ester,methyl pyridine-2-acetate
74264
methyl 2-pyridin-2-ylacetate
COC(=O)CC1=CC=CC=N1
1658-42-0
C8H9NO2
151.165
MFCD00006358
ORAKNQSHWMHCEY-UHFFFAOYSA-N
methyl 2-pyridylacetate,methyl 2-pyridin-2-yl acetate,2-pyridylacetic acid methyl ester,2-pyridineacetic acid, methyl ester,methyl 2-pyridinylacetate,methyl pyridin-2-ylacetate,methyl 2-pyridineacetate,2-pyridineacetic acid methyl ester,pyridin-2-yl-acetic acid methyl ester,methyl pyridine-2-acetate
74264
methyl 2-pyridin-2-ylacetate
COC(=O)CC1=CC=CC=N1
Refractive Index 1.5050 to 1.5070 (20°C, 589nm)
Color Yellow
Packaging Glass bottle
Quantity 5g
Density 1.119g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (C5H4N)CH2COOCH3
Formula Weight 151.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Methyl 2-pyridylacetate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.