Alfa Aesar™ Methyl 2-thiopheneacetate, 98%

Manufacturer: Alfa Aesar™ A1211006

Quantity 5g

 View more versions of this product

Catalog No. AAA1211006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19432-68-9
    Molecular Formula C7H8O2S
    Formula Weight 156.2
    MDL Number MFCD00041213
    Beilstein 120622
    Synonym Methyl 2-thienylacetate; 2-Thiopheneacetic acid methyl ester
    Chemical Name or Material Methyl 2-thiopheneacetate

    Specifications

    Formula Weight 156.2
    Molecular Formula C7H8O2S
    MDL Number MFCD00041213
    Quantity 5g
    CAS 19432-68-9
    Beilstein 120622
    Boiling Point 100° to 104°C (14mmHg)
    Synonym Methyl 2-thienylacetate; 2-Thiopheneacetic acid methyl ester
    Chemical Name or Material Methyl 2-thiopheneacetate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit