Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, 98%

Click to view available options
Quantity:
5g
25g
100g
200573-11-1
C13H9F3O5S
334.265
MFCD04039773
BDXDWDCWSCPFEW-UHFFFAOYSA-N
methyl 2-trifluoromethylsulfonyloxy-1-naphthoate,methyl 2-trifluoromethanesulfonyloxy-1-naphthoate,methyl 2-trifluoromethyl sulfonyl oxy-1-naphthoate,methyl 2-trifluoromethanesulfonyloxy naphthalene-1-carboxylate,acmc-20aki5,methyl 2-trifluoromethylsulfonyloxy naphthalene-1-carboxylate,1-carbomethoxy-2-trifluoromethylsulfonyloxy naphthalene,2-trifluoromethylsulfonyloxy-1-naphthoic acid methyl ester,2-trifluoromethylsulfonyloxy naphthalene-1-carboxylic acid methyl ester
11034997
methyl 2-(trifluoromethylsulfonyloxy)naphthalene-1-carboxylate
COC(=O)C1=C(C=CC2=CC=CC=C21)OS(=O)(=O)C(F)(F)F
200573-11-1
C13H9F3O5S
334.265
MFCD04039773
BDXDWDCWSCPFEW-UHFFFAOYSA-N
methyl 2-trifluoromethylsulfonyloxy-1-naphthoate,methyl 2-trifluoromethanesulfonyloxy-1-naphthoate,methyl 2-trifluoromethyl sulfonyl oxy-1-naphthoate,methyl 2-trifluoromethanesulfonyloxy naphthalene-1-carboxylate,acmc-20aki5,methyl 2-trifluoromethylsulfonyloxy naphthalene-1-carboxylate,1-carbomethoxy-2-trifluoromethylsulfonyloxy naphthalene,2-trifluoromethylsulfonyloxy-1-naphthoic acid methyl ester,2-trifluoromethylsulfonyloxy naphthalene-1-carboxylic acid methyl ester
11034997
methyl 2-(trifluoromethylsulfonyloxy)naphthalene-1-carboxylate
COC(=O)C1=C(C=CC2=CC=CC=C21)OS(=O)(=O)C(F)(F)F
Quantity 100g
Melting Point 41°C to 43°C
Formula Weight 334.27
Percent Purity 98%
Chemical Name or Material Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning