Alfa Aesar™ Methyl 3-[3-(aminomethyl)phenyl]propionate, 99%

Manufacturer: Alfa Aesar™ H5094103

Quantity 1g

 View more versions of this product

Catalog No. AAH5094103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100511-83-9
    Molecular Formula C11H15NO2
    Formula Weight 193.25
    MDL Number MFCD08064274
    Synonym 3-[3-(Aminomethyl)phenyl]propionic acid methyl ester
    Chemical Name or Material Methyl 3-[3-(aminomethyl)phenyl]propionate

    Specifications

    Formula Weight 193.25
    Synonym 3-[3-(Aminomethyl)phenyl]propionic acid methyl ester
    Quantity 1g
    Molecular Formula C11H15NO2
    MDL Number MFCD08064274
    CAS 100511-83-9
    Chemical Name or Material Methyl 3-[3-(aminomethyl)phenyl]propionate
    Assay 99%

    Description

    Applications
    Methyl 3-[3-(aminomethyl)phenyl]propionate is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit