Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,4,5-trimethoxybenzoate, 98+%

Catalog No. aaa1193314
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 1916-07-0 | C11H14O5 | 226.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1916-07-0
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00008431
InChI Key KACHFMOHOPLTNX-UHFFFAOYSA-N
Synonym trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8
PubChem CID 15956
IUPAC Name methyl 3,4,5-trimethoxybenzoate
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)OC

Specifications

Melting Point 81°C to 85°C
Boiling Point 274°C to 275°C
Quantity 25 g
Beilstein 2218156
Formula Weight 226.23
Percent Purity ≥98%
Chemical Name or Material Methyl 3,4,5-trimethoxybenzoate
EINECSNumber 217-629-3
RTECSNumber DI0820000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.