Alfa Aesar™ Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride, 97%

Manufacturer: Alfa Aesar™ H5103103

Quantity 1g

 View more versions of this product

Catalog No. AAH5103103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 103565-40-8
    Molecular Formula C11H15NO2·HCl
    Formula Weight 229.71
    MDL Number MFCD11113035
    Beilstein 14326402
    Synonym 3-[4-(Aminomethyl)phenyl]propionic acid methyl ester hydrochloride
    Chemical Name or Material Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride

    Specifications

    Formula Weight 229.71
    Molecular Formula C11H15NO2·HCl
    MDL Number MFCD11113035
    Synonym 3-[4-(Aminomethyl)phenyl]propionic acid methyl ester hydrochloride
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 103565-40-8
    Beilstein 14326402
    Health Hazard 1 Warning
    Chemical Name or Material Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit