Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,4-dichlorobenzo[b]thiophene-2-carboxylate, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
444905-19-5
C10H6Cl2O2S
261.116
MFCD02056432
SKWQDBQCLUXTKF-UHFFFAOYSA-N
methyl 3,4-dichlorobenzo b thiophene-2-carboxylate,enamine_005470,benzo b thiophene-2-carboxylic acid, 3,4-dichloro-, methyl ester
2181054
methyl 3,4-dichloro-1-benzothiophene-2-carboxylate
COC(=O)C1=C(C2=C(S1)C=CC=C2Cl)Cl
444905-19-5
C10H6Cl2O2S
261.116
MFCD02056432
SKWQDBQCLUXTKF-UHFFFAOYSA-N
methyl 3,4-dichlorobenzo b thiophene-2-carboxylate,enamine_005470,benzo b thiophene-2-carboxylic acid, 3,4-dichloro-, methyl ester
2181054
methyl 3,4-dichloro-1-benzothiophene-2-carboxylate
COC(=O)C1=C(C2=C(S1)C=CC=C2Cl)Cl
Quantity 1g
Melting Point 116°C to 118°C
Formula Weight 261.12
Percent Purity 97%
Chemical Name or Material Methyl 3,4-dichlorobenzo[b]thiophene-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures