Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3,4-dichlorophenylacetate, 97+%

Click to view available options
Quantity:
2g
10g
50g
6725-44-6
C9H8Cl2O2
219.061
MFCD00800680
FWVOAIHAIWHHDM-UHFFFAOYSA-N
methyl 2-3,4-dichlorophenyl acetate,methyl 3,4-dichlorophenylacetate,3,4-dichlorophenylacetic acid methyl ester,3,4-dichloro-phenyl-acetic acid methyl ester,benzeneacetic acid, 3,4-dichloro-, methyl ester,pubchem20251,acmc-1bhwn,methyl 3,4-dichloro-phenylacetate,methyl 2-3,4-dichlorophenyl ethanoate,3,4 dichlorophenyl acetic acid methyl ester
1268257
methyl 2-(3,4-dichlorophenyl)acetate
COC(=O)CC1=CC(=C(C=C1)Cl)Cl
6725-44-6
C9H8Cl2O2
219.061
MFCD00800680
FWVOAIHAIWHHDM-UHFFFAOYSA-N
methyl 2-3,4-dichlorophenyl acetate,methyl 3,4-dichlorophenylacetate,3,4-dichlorophenylacetic acid methyl ester,3,4-dichloro-phenyl-acetic acid methyl ester,benzeneacetic acid, 3,4-dichloro-, methyl ester,pubchem20251,acmc-1bhwn,methyl 3,4-dichloro-phenylacetate,methyl 2-3,4-dichlorophenyl ethanoate,3,4 dichlorophenyl acetic acid methyl ester
1268257
methyl 2-(3,4-dichlorophenyl)acetate
COC(=O)CC1=CC(=C(C=C1)Cl)Cl
Quantity 10g
Melting Point 27°C to 30°C
Formula Weight 219.07
Percent Purity ≥97%
Chemical Name or Material Methyl 3,4-dichlorophenylacetate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning