Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,4-dichlorophenylacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6725-44-6
C9H8Cl2O2
219.061
MFCD00800680
FWVOAIHAIWHHDM-UHFFFAOYSA-N
methyl 2-3,4-dichlorophenyl acetate,methyl 3,4-dichlorophenylacetate,3,4-dichlorophenylacetic acid methyl ester,3,4-dichloro-phenyl-acetic acid methyl ester,benzeneacetic acid, 3,4-dichloro-, methyl ester,pubchem20251,acmc-1bhwn,methyl 3,4-dichloro-phenylacetate,methyl 2-3,4-dichlorophenyl ethanoate,3,4 dichlorophenyl acetic acid methyl ester
1268257
methyl 2-(3,4-dichlorophenyl)acetate
COC(=O)CC1=CC(=C(C=C1)Cl)Cl
6725-44-6
C9H8Cl2O2
219.061
MFCD00800680
FWVOAIHAIWHHDM-UHFFFAOYSA-N
methyl 2-3,4-dichlorophenyl acetate,methyl 3,4-dichlorophenylacetate,3,4-dichlorophenylacetic acid methyl ester,3,4-dichloro-phenyl-acetic acid methyl ester,benzeneacetic acid, 3,4-dichloro-, methyl ester,pubchem20251,acmc-1bhwn,methyl 3,4-dichloro-phenylacetate,methyl 2-3,4-dichlorophenyl ethanoate,3,4 dichlorophenyl acetic acid methyl ester
1268257
methyl 2-(3,4-dichlorophenyl)acetate
COC(=O)CC1=CC(=C(C=C1)Cl)Cl
Quantity 5g
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
Assay Percent Range 99%
Formula Weight 219.07
Physical Form Liquid After Melting
Percent Purity ≥98.5%
Chemical Name or Material Methyl 3, 4-dichlorophenylacetate, 99%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.