Alfa Aesar™ Methyl 3,4-dimethoxybenzoate, 98+%

Manufacturer: Alfa Aesar™ B2356830

Quantity 250g

 View more versions of this product

Catalog No. AAB2356830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2150-38-1
    Molecular Formula C10H12O4
    Formula Weight 196.2
    MDL Number MFCD00008430
    Beilstein 2215244
    Synonym 3,4-Dimethoxybenzoic acid methyl ester; Methyl veratrate
    Chemical Name or Material Methyl 3,4-dimethoxybenzoate

    Specifications

    Formula Weight 196.2
    TSCA TSCA
    Molecular Formula C10H12O4
    MDL Number MFCD00008430
    CAS 2150-38-1
    Beilstein 2215244
    Synonym 3,4-Dimethoxybenzoic acid methyl ester; Methyl veratrate
    Melting Point 57° to 61°C
    Boiling Point 283°C
    Quantity 250g
    Chemical Name or Material Methyl 3,4-dimethoxybenzoate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit