Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,4-dimethoxybenzoate, 98+%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Plastic bottle
2150-38-1
C10H12O4
196.2
MFCD00008430
BIGQPYZPEWAPBG-UHFFFAOYSA-N
methyl veratrate,benzoic acid, 3,4-dimethoxy-, methyl ester,3,4-dimethoxybenzoic acid methyl ester,veratric acid, methyl ester,methylveratrate,pubchem13009,acmc-1cfay,veratric acid methyl ester,methyl 3,4 dimethoxybenzoate,methyl 3, 4-dimethoxybenzoate
16522
CHEBI:86906
methyl 3,4-dimethoxybenzoate
COC1=C(C=C(C=C1)C(=O)OC)OC
2150-38-1
C10H12O4
196.2
MFCD00008430
BIGQPYZPEWAPBG-UHFFFAOYSA-N
methyl veratrate,benzoic acid, 3,4-dimethoxy-, methyl ester,3,4-dimethoxybenzoic acid methyl ester,veratric acid, methyl ester,methylveratrate,pubchem13009,acmc-1cfay,veratric acid methyl ester,methyl 3,4 dimethoxybenzoate,methyl 3, 4-dimethoxybenzoate
16522
CHEBI:86906
methyl 3,4-dimethoxybenzoate
COC1=C(C=C(C=C1)C(=O)OC)OC
Boiling Point 283°C
Packaging Plastic bottle
Quantity 50g
Infrared Spectrum Authentic
Color Brown
Melting Point 58°C to 62°C
Assay Percent Range 98% min. (GC)
Linear Formula (CH3O)2C6H3CO2CH3
Beilstein 10, 396
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol,acetone
Formula Weight 196.2
Physical Form Granular Powder
Percent Purity 98+%
Chemical Name or Material Methyl 3, 4-dimethoxybenzoate, 98+%
Show More Show Less

EINECSNumber : 218-424-1

TSCA : TSCA