Alfa Aesar™ Methyl 3-(4-methyl-1-piperazinyl)benzoate, 97%

Manufacturer: Alfa Aesar™ H5577006

Quantity 5g

 View more versions of this product

Catalog No. AAH5577006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 474334-89-9
    Formula Weight 234.3
    MDL Number MFCD07369792
    Synonym 1-(3-Carbomethoxyphenyl)-4-methylpiperazine
    Chemical Name or Material Methyl 3-(4-methyl-1-piperazinyl)benzoate

    Specifications

    Formula Weight 234.3
    Boiling Point 169° to 172°C (0.9mmHg)
    Refractive Index 1.5630
    Physical Form Liquid
    Color Colorless to dark yellow
    Synonym 1-(3-Carbomethoxyphenyl)-4-methylpiperazine
    Density 1.119g/mL
    MDL Number MFCD07369792
    CAS 474334-89-9
    Quantity 5g
    Chemical Name or Material Methyl 3-(4-methyl-1-piperazinyl)benzoate
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit