Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3,5-dibromo-2-hydroxybenzoate, 98%

Click to view available options
Quantity:
25g
100g
500g
21702-79-4
C8H6Br2O3
309.941
MFCD00017534
KISQISIKTRRMOX-UHFFFAOYSA-N
methyl 3,5-dibromosalicylate,benzoic acid, 3,5-dibromo-2-hydroxy-, methyl ester,3,5-dibromo-2-hydroxybenzoic acid methyl ester,pubchem3976,acmc-1cpm5,rarechem al bf 0030,timtec-bb sbb001273,3,5-dibromo-2-hydroxymethyl benzoate,benzoic acid,3,5-dibromo-2-hydroxy-, methyl ester
624217
methyl 3,5-dibromo-2-hydroxybenzoate
COC(=O)C1=CC(=CC(=C1O)Br)Br
21702-79-4
C8H6Br2O3
309.941
MFCD00017534
KISQISIKTRRMOX-UHFFFAOYSA-N
methyl 3,5-dibromosalicylate,benzoic acid, 3,5-dibromo-2-hydroxy-, methyl ester,3,5-dibromo-2-hydroxybenzoic acid methyl ester,pubchem3976,acmc-1cpm5,rarechem al bf 0030,timtec-bb sbb001273,3,5-dibromo-2-hydroxymethyl benzoate,benzoic acid,3,5-dibromo-2-hydroxy-, methyl ester
624217
methyl 3,5-dibromo-2-hydroxybenzoate
COC(=O)C1=CC(=CC(=C1O)Br)Br
Melting Point 150°C to 152°C
Quantity 500g
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-2-hydroxybenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning