Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,5-dibromo-4-hydroxybenzoate, 99%, Acros Organics

Click to view available options
Quantity:
5g
41727-47-3
C8H6Br2O3
309.941
MFCD00016416
NVGJGYZKXBLIKY-UHFFFAOYSA-N
3,5-dibromo-4-hydroxybenzoic acid methyl ester,3,5-dibromo-4-hydroxy-benzoic acid methyl ester,methyl 3,5-bis bromanyl-4-oxidanyl-benzoate,acmc-20akhs,pubchem3978,methyl dibromosalicylate,rarechem al bf 0231,timtec-bb sbb012530,labotest-bb lt00455670,labotest-bb lt03332530
726975
methyl 3,5-dibromo-4-hydroxybenzoate
COC(=O)C1=CC(=C(C(=C1)Br)O)Br
41727-47-3
C8H6Br2O3
309.941
MFCD00016416
NVGJGYZKXBLIKY-UHFFFAOYSA-N
3,5-dibromo-4-hydroxybenzoic acid methyl ester,3,5-dibromo-4-hydroxy-benzoic acid methyl ester,methyl 3,5-bis bromanyl-4-oxidanyl-benzoate,acmc-20akhs,pubchem3978,methyl dibromosalicylate,rarechem al bf 0231,timtec-bb sbb012530,labotest-bb lt00455670,labotest-bb lt03332530
726975
methyl 3,5-dibromo-4-hydroxybenzoate
COC(=O)C1=CC(=C(C(=C1)Br)O)Br
Quantity 5g
Color White
Melting Point 122°C to 125°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Formula Weight 309.94
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material Methyl 3, 5-dibromo-4-hydroxybenzoate, 99%

EINECSNumber : 255-521-8