Alfa Aesar™ Methyl 3,5-dibromo-4-methylbenzoate, 98%

Manufacturer: Alfa Aesar™ B2534806

Quantity 5g

 View more versions of this product

Catalog No. AAB2534806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74896-66-5
    Molecular Formula C9H8Br2O2
    Formula Weight 307.98
    MDL Number MFCD00010729
    Beilstein 2580945
    Synonym 3,5-Dibromo-4-methylbenzoic acid methyl ester
    Chemical Name or Material Methyl 3,5-dibromo-4-methylbenzoate

    Specifications

    Formula Weight 307.98
    Melting Point 86° to 90°C
    Quantity 5g
    Molecular Formula C9H8Br2O2
    MDL Number MFCD00010729
    CAS 74896-66-5
    Beilstein 2580945
    Synonym 3,5-Dibromo-4-methylbenzoic acid methyl ester
    Chemical Name or Material Methyl 3,5-dibromo-4-methylbenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit