Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3,5-dimethoxybenzoate, 98%

Manufacturer:  Alfa Aesar™ A1148618

Quantity 50g

 View more versions of this product

Catalog No. AAA1148618


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2150-37-0
196.2
2693126
Methyl 3,5-dimethoxybenzoate
C10H12O4
MFCD00008432
3,5-Dimethoxybenzoic acid methyl ester

Specifications

2150-37-0
196.2
2693126
40° to 43°C
50g
Methyl 3,5-dimethoxybenzoate
C10H12O4
MFCD00008432
3,5-Dimethoxybenzoic acid methyl ester
298°C
>110°C (230°F)
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit