Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3,5-dimethylbenzoate, 98%

Manufacturer:  Alfa Aesar™ A1228706

Quantity 5g

 View more versions of this product

Catalog No. AAA1228706


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

25081-39-4
164.21
3,5-Dimethylbenzoic acid methyl ester
C10H12O2
MFCD00077971
Methyl 3,5-dimethylbenzoate

Specifications

25081-39-4
164.21
3,5-Dimethylbenzoic acid methyl ester
239° to 240°C
106°C (224°F)
98%
C10H12O2
MFCD00077971
31° to 33°C
5g
Methyl 3,5-dimethylbenzoate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit