Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3,5-dinitrobenzoate, 98%

Click to view available options
Quantity:
10g
25g
100g
250g
2702-58-1
C8H6N2O6
226.144
MFCD00017016
POGCCFLNFPIIGW-UHFFFAOYSA-N
benzoic acid, 3,5-dinitro-, methyl ester,3,5-dinitro-benzoic acid methyl ester,methyl 3,5-dinitrobenzoate,methyl-3,5-dinitrobenzoate,acmc-1cr6q,d0x5an,ksc494q3h,methyl-3,5-dinitro benzoate,3,5-dinitrobenzoic acid methyl ester
222067
methyl 3,5-dinitrobenzoate
COC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]
2702-58-1
C8H6N2O6
226.144
MFCD00017016
POGCCFLNFPIIGW-UHFFFAOYSA-N
benzoic acid, 3,5-dinitro-, methyl ester,3,5-dinitro-benzoic acid methyl ester,methyl 3,5-dinitrobenzoate,methyl-3,5-dinitrobenzoate,acmc-1cr6q,d0x5an,ksc494q3h,methyl-3,5-dinitro benzoate,3,5-dinitrobenzoic acid methyl ester
222067
methyl 3,5-dinitrobenzoate
COC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]
Melting Point 108°C to 112°C
Quantity 250g
Beilstein 1252593
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dinitrobenzoate