Alfa Aesar™ Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate, 97%

Manufacturer: Alfa Aesar™ H2623303

Quantity 1g

 View more versions of this product

Catalog No. AAH2623303

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 129332-45-2
    Molecular Formula C8H8N2O2S2
    Formula Weight 228.28
    MDL Number MFCD00052752
    UN Number UN3439
    Synonym 3-Amino-4-cyano-5-(methylthio)thiophene-2-carboxylic acid methyl ester
    Chemical Name or Material Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate

    Specifications

    Formula Weight 228.28
    Molecular Formula C8H8N2O2S2
    MDL Number MFCD00052752
    CAS 129332-45-2
    UN Number UN3439
    Synonym 3-Amino-4-cyano-5-(methylthio)thiophene-2-carboxylic acid methyl ester
    Health Hazard 1 Danger
    Melting Point 205° to 208°C
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate
    Assay 97%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit