Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-amino-4-hydroxybenzoate, 95%

Click to view available options
Quantity:
1g
5g
536-25-4
C8H9NO3
167.164
MFCD06659667
VNQABZCSYCTZMS-UHFFFAOYSA-N
orthocaine,orthoform,aminobenz,orthoderm,3-amino-4-hydroxybenzoic acid methyl ester,benzoic acid, 3-amino-4-hydroxy-, methyl ester,methyl m-amino-p-hydroxybenzoate,methyl 4-hydroxy-3-aminobenzoate,methyl 3-amino-4-hydroxybenzenecarboxylate,unii-jk16yi13qk
10815
CHEBI:34904
methyl 3-amino-4-hydroxybenzoate
COC(=O)C1=CC(=C(C=C1)O)N
536-25-4
C8H9NO3
167.164
MFCD06659667
VNQABZCSYCTZMS-UHFFFAOYSA-N
orthocaine,orthoform,aminobenz,orthoderm,3-amino-4-hydroxybenzoic acid methyl ester,benzoic acid, 3-amino-4-hydroxy-, methyl ester,methyl m-amino-p-hydroxybenzoate,methyl 4-hydroxy-3-aminobenzoate,methyl 3-amino-4-hydroxybenzenecarboxylate,unii-jk16yi13qk
10815
CHEBI:34904
methyl 3-amino-4-hydroxybenzoate
COC(=O)C1=CC(=C(C=C1)O)N
Quantity 1g
Melting Point 141°C to 143°C
Beilstein 5424619
Formula Weight 167.16
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methyl 3-amino-4-hydroxybenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG2625000

TSCA : TSCA