Alfa Aesar™ Methyl 3-amino-4-methoxybenzoate, 97%

Manufacturer: Alfa Aesar™ A1205214

Quantity 25g

 View more versions of this product

Catalog No. AAA1205214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24812-90-6
    Molecular Formula C9H11NO3
    Formula Weight 181.19
    MDL Number MFCD00017196
    Beilstein 2968002
    Synonym 3-Amino-4-methoxybenzoic acid methyl ester
    Chemical Name or Material Methyl 3-amino-4-methoxybenzoate

    Specifications

    Formula Weight 181.19
    CAS 24812-90-6
    MDL Number MFCD00017196
    Beilstein 2968002
    Synonym 3-Amino-4-methoxybenzoic acid methyl ester
    Melting Point 84° to 86°C
    Molecular Formula C9H11NO3
    Quantity 25g
    Chemical Name or Material Methyl 3-amino-4-methoxybenzoate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit