Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
85006-31-1
C7H9NO2S
171.22
YICRPERKKBDRSP-UHFFFAOYSA-N
methyl3-amino-4-methylthiophene-2-carboxylate,3-amino-4-methylthiophene-2-carboxylic acid methyl ester,unii-3la24473as,3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester,2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester,methyl 3-amino-4-methyl-2-thenoate,methyl 3-amino-4-methyl-2-thiophenecarboxylate,methyl-3-amino-4-methylthiophene-2-carboxylate,methyl 3-amino-4-methyl-thiophene-2-carboxylate,3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
123584
methyl 3-amino-4-methylthiophene-2-carboxylate
CC1=CSC(=C1N)C(=O)OC
85006-31-1
C7H9NO2S
171.22
YICRPERKKBDRSP-UHFFFAOYSA-N
methyl3-amino-4-methylthiophene-2-carboxylate,3-amino-4-methylthiophene-2-carboxylic acid methyl ester,unii-3la24473as,3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester,2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester,methyl 3-amino-4-methyl-2-thenoate,methyl 3-amino-4-methyl-2-thiophenecarboxylate,methyl-3-amino-4-methylthiophene-2-carboxylate,methyl 3-amino-4-methyl-thiophene-2-carboxylate,3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
123584
methyl 3-amino-4-methylthiophene-2-carboxylate
CC1=CSC(=C1N)C(=O)OC
Flash Point >100°C
Melting Point 85°C to 88°C
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 171.22
Percent Purity 98%
Chemical Name or Material Methyl 3-amino-4-methyl-2-thiophenecarboxylate

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

EINECSNumber : 285-060-8